Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-HSP60 Goat Polyclonal Antibody
Anti-HSP60 Goat Polyclonal Antibody
Supplier:  ENZO LIFE SCIENCES
Certificates
Anti-HSP60 Goat Polyclonal Antibody
Supplier:  ENZO LIFE SCIENCES

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Catalog No:
  • ENZOADISPA828J
  • ENZOADISPA828F
  • ENZOADISPA828D
  • Antigen symbol:
  • HSP60
  • HSP60
  • HSP60
  • Antigen name:
  • Heat Shock Protein 60
  • Heat Shock Protein 60
  • Heat Shock Protein 60
  • Antigen synonyms:
  • Mytonin Chaperone 60|GROEL|HSP65|P60 lymphocyte protein|HLD4|HSP-60|HuCHA60|mitochondrial matrix protein P1|chaperonin 60|HSP60|mitochondrial|CPN60|short heat shock protein 60 Hsp60s1|heat shock protein 65|60 kDa chaperonin|60 kDa heat shock protein|SPG13
  • Mytonin Chaperone 60|GROEL|HSP65|P60 lymphocyte protein|HLD4|HSP-60|HuCHA60|mitochondrial matrix protein P1|chaperonin 60|HSP60|mitochondrial|CPN60|short heat shock protein 60 Hsp60s1|heat shock protein 65|60 kDa chaperonin|60 kDa heat shock protein|SPG13
  • Mytonin Chaperone 60|GROEL|HSP65|P60 lymphocyte protein|HLD4|HSP-60|HuCHA60|mitochondrial matrix protein P1|chaperonin 60|HSP60|mitochondrial|CPN60|short heat shock protein 60 Hsp60s1|heat shock protein 65|60 kDa chaperonin|60 kDa heat shock protein|SPG13
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Polyclonal
  • Reactivity:
  • Human
  • Host:
  • Goat
  • Goat
  • Goat
  • Western blot:
  • Yes
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Yes
  • Form:
  • Liquid, in PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide
  • Liquid, in PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide
  • Liquid, in PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide
  • Purification:
  • Protein G affinity purified
  • Protein G affinity purified
  • Protein G affinity purified

Specifications

Frequently Bought Together