Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-CRP55 Rabbit Polyclonal Antibody
Anti-CRP55 Rabbit Polyclonal Antibody
Supplier:  ENZO LIFE SCIENCES
Certificates
Anti-CRP55 Rabbit Polyclonal Antibody
Supplier:  ENZO LIFE SCIENCES

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Catalog No:
  • ENZOADISPA600D
  • ENZOADISPA600J
  • ENZOADISPA600F
  • Antigen symbol:
  • CRP55
  • CRP55
  • CRP55
  • Antigen name:
  • Calreticulin
  • Calreticulin
  • Calreticulin
  • Antigen synonyms:
  • CRT|FLJ26680|CALR|cC1qR|SSA|endoplasmic reticulum resident protein 60|HEL-S-99n|RO
  • CRT|FLJ26680|CALR|cC1qR|SSA|endoplasmic reticulum resident protein 60|HEL-S-99n|RO
  • CRT|FLJ26680|CALR|cC1qR|SSA|endoplasmic reticulum resident protein 60|HEL-S-99n|RO
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Polyclonal
  • Reactivity:
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Rabbit
  • Western blot:
  • Yes
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Yes
  • Form:
  • Liquid. In PBS containing 50% glycerol and 0,09% sodium azide.
  • Liquid. In PBS containing 50% glycerol and 0,09% sodium azide.
  • Liquid. In PBS containing 50% glycerol and 0,09% sodium azide.
  • Purification:
  • Protein A affinity purified
  • Protein A affinity purified
  • Protein A affinity purified

Specifications

Frequently Bought Together