Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-HMOX1 Mouse Monoclonal Antibody
Anti-HMOX1 Mouse Monoclonal Antibody
Supplier:  ENZO LIFE SCIENCES
Certificates
Anti-HMOX1 Mouse Monoclonal Antibody
Supplier:  ENZO LIFE SCIENCES

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Catalog No:
  • ENZOADIOSA111J
  • ENZOADIOSA111F
  • ENZOADIOSA111D
  • Antigen symbol:
  • HMOX1
  • HMOX1
  • HMOX1
  • Antigen name:
  • Heme Oxygenase 1
  • Heme Oxygenase 1
  • Heme Oxygenase 1
  • Antigen synonyms:
  • Hsp32|heat shock protein 32|HMOX1|HO-1|heme orygenase 1
  • Hsp32|heat shock protein 32|HMOX1|HO-1|heme orygenase 1
  • Hsp32|heat shock protein 32|HMOX1|HO-1|heme orygenase 1
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Reactivity:
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Isotype:
  • IgG2b
  • IgG2b
  • IgG2b
  • Western blot:
  • Yes
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Form:
  • Liquid. In PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide.
  • Liquid. In PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide.
  • Liquid. In PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide.
  • Purification:
  • Protein G affinity purified
  • Protein G affinity purified
  • Protein G affinity purified

Specifications

Frequently Bought Together