Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-GSK3A Mouse Monoclonal Antibody [clone: 6D3]
Anti-GSK3A Mouse Monoclonal Antibody [clone: 6D3]
Catalog # ENZOADI905762100
Supplier:  ENZO LIFE SCIENCES
Anti-GSK3A Mouse Monoclonal Antibody [clone: 6D3]
Catalog # ENZOADI905762100
Supplier:  ENZO LIFE SCIENCES
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glycogen Synthase Kinase 3 alpha
  • Antigen symbol:
    GSK3A
  • Clonality:
    Monoclonal
  • Clone:
    6D3
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Lyophilized from 1 ml of 2× PBS containing 0,09% sodium azide, PEG, and sucrose.
  • Antigen synonyms:
    GSK-3 alpha|Glycogen synthase kinase-3 alpha|GSK3A|Serine/threonine-protein kinase GSK3A
  • Purification:
    Thiophilic adsorption and size exclusion chromatography purified
  • Pk:
    100 µG

Specifications

About this item

Glycogen Synthase Kinase 3β (GSK-3β) is a unique serine/threonine kinase that is inactivated by phosphorylation. In response to insulin binding, PKB/AKT phosphorylates GSK-3β on serine 9, which prevents GSK-3β from phosphorylating glycogen synthase. Unphosphorylated glycogen synthase is active and able to synthesize glycogen. GSK-3β is also unique in that it requires a substrate that has been phosphorylated by a distinct kinase before it can phosphorylate the substrate. This phosphate priming mechanism explains why phosphorylation of serine 9 inactivates GSK-3β. The phosphorylated serine binds to the GSK-3β priming phosphate position and prevents binding of alternative substrates. In addition to insulin signaling, GSK-3β participates in the Wnt signaling pathway, where it forms a complex with axin, beta-catenin and adenomatous polyposis coli (APC) protein. In the presence of Wnts, GSK-3β is unable to phosphorylate beta-catenin, which leads to stabilization of beta-catenin. The Wnt pathway inactivates GSK-3β via the proteins, Dishevelled and FRAT, which disrupt the interaction of GSK-3β with axin, beta-catenin, and APC. Clinically, there is considerable interest in GSK-3β inhibitors because they may mimic the effect of insulin or reduce the hyperphosphorylation of Tau that is observed in Alzheimer's Disease.

  • For Western Blot

Recognizes human and mouse GSK-3alpha/beta.

Reconstitute with 1ml water (15 minutes at room temperature).

Type: Primary
Antigen: GSK-3ALPHA
Clonality: Monoclonal
Clone: 6D3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse