Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Glutathione-S-Transferase
- Antigen symbol:GST
- Clonality:Monoclonal
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Western blot:Yes
- Seq:02
- Form:Liquid. In PBS containing 0,01% sodium azide.
- Antigen synonyms:GSTP1-1|PI|DFN7|GST3|GSTP1|FAEES3
- Purification:Protein G affinity purified
- Pk:100 µG
Specifications
About this item
The GST superfamily of enzymes play an important role in detoxification of xenobiotics by catalyzing the conjugation of many hydrophobic and electrophilic compounds with reduced glutathione. There are four main GST classes; alpha, mu, pi, and tau. GST-P is a member of the pi class of GSTs, and its expression is specifically increased in the early stages of rat hepatocarcinogenesis. Due to its high affinity for glutathione, GST is frequently utilized as an approximately 26 kDa fusion tag for the expression and purification of recombinant proteins.
- For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot
Species independent
Type: Primary
Antigen: GST
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Species Independent