
Sulphuric acid ≥98% for the determination of nitrogen
Supplier: PanReac AppliChem
Certificates
About this item
CAS Number:7664-93-9
:
Formula:H₂SO₄
MDL Number:MFCD00064589
:98.08 g/mol
Boiling Pt:330 °C
Melting Pt:–15 °C
Density:1,84
:GHS05
Specification Test Results
- Assay:98%
- Appearence:Clear colorless liquid
- Density 20/4:1,84
- Residue on ignition:0,0005%
- Nitrogen compounds (as N):0,0005%
Specification Test Results
Frequently Bought Together