Order Entry
Northern Ireland
ContactUsLinkComponent
Rack for cuvettes
Rack for cuvettes
Catalog # 634-8100
Supplier:  Brand
undefined
Rack for cuvettes
Catalog # 634-8100
Supplier:  Brand

Specifications

  • For:
    16 (2×8)
  • W×D×H:
    210×70×38 mm
  • Pk:
    1

Specifications

About this item

PP.

  • Numbered positions to aid sample identification
  • Autoclavable at 121 °C

Rack with numbered positions for 2×8 cuvettes of 10 mm path length.