To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen symbol:TRPC5
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Polyclonal antibody for TRPC5 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. TRPC5 information: Molecular Weight: 111412 MW; Subcellular Localization: Cell membrane ; Multi-pass membrane protein ; Tissue Specificity: Expressed in brain with higher levels in fetal brain. Found in cerebellum and occipital pole.
Type: Primary
Antigen: TRPC5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human