Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:TNF Beta
- Antigen symbol:TNFB
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Polyclonal antibody for TNF BETA/LTA detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. TNF BETA/LTA information: Molecular Weight: 22297 MW; Subcellular Localization: Secreted. Membrane. The homotrimer is secreted. The heterotrimer is membrane-associated.
Type: Primary
Antigen: TNF Beta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human