Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Src Polyclonal Antibody
Anti-Src Polyclonal Antibody
Catalog # BSBTPB9059
Supplier:  Boster Bio
undefined
Anti-Src Polyclonal Antibody
Catalog # BSBTPB9059
Supplier:  Boster Bio
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Src
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Polyclonal antibody for SRC detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. SRC information: Molecular Weight: 59835 MW; Subcellular Localization: Cell membrane. Mitochondrion inner membrane. Nucleus. Cytoplasm, cytoskeleton. Localizes to focal adhesion sites following integrin engagement. Localization to focal adhesion sites requires myristoylation and the SH3 domain; Tissue Specificity: Expressed ubiquitously. Platelets, neurons and osteoclasts express 5-fold to 200-fold higher levels than most other tissues.

Type: Primary
Antigen: Src
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human