Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen symbol:RACK1
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for GNB2L1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. GNB2L1 information: Molecular Weight: 35077 MW; Subcellular Localization: Cell membrane; Peripheral membrane protein. Cytoplasm. Cytoplasm, perinuclear region. Cytoplasm, cytoskeleton. Nucleus. Perikaryon . Cell projection, dendrite . Cell projection, phagocytic cup. Recruited to the plasma membrane through interaction with KRT1 which binds to membrane-bound ITGB1. Also associated with the membrane in oncogene-transformed cells. PKC activation induces translocation from the perinuclear region to the cell periphery. In the brain, detected mainly in cell bodies and dendrites with little expression in axonal fibers or nuclei. Localized to phagocytic cups following infection by Y.pestis; Tissue Specificity: In the liver, expressed at higher levels in activated hepatic stellate cells than in hepatocytes or Kupffer cells. Up-regulated in hepatocellular carcinomas and in the adjacent non-tumor liver tissue.
Type: Primary
Antigen: RACK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat