Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-PRX Polyclonal Antibody
Anti-PRX Polyclonal Antibody
Catalog # BSBTA01686
Supplier:  Boster Bio
undefined
Anti-PRX Polyclonal Antibody
Catalog # BSBTA01686
Supplier:  Boster Bio

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    PRX
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for PRX detection. Tested with WB, IHC-F, ICC, FCM, Direct ELISA in Human;Mouse;Rat.

Type: Primary
Antigen: PRX
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat