Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-GST3/GST Pi Polyclonal Antibody
Anti-GST3/GST Pi Polyclonal Antibody
Catalog # BSBTPB9184
Supplier:  Boster Bio
undefined
Anti-GST3/GST Pi Polyclonal Antibody
Catalog # BSBTPB9184
Supplier:  Boster Bio

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    GST3/GST Pi
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Polyclonal antibody for GST PI/GSTP1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. GST PI/GSTP1 information: Molecular Weight: 23356 MW; Subcellular Localization: Cytoplasm . Mitochondrion . Nucleus . The 83 N-terminal amino acids function as un uncleaved transit peptide, and arginine residues within it are crucial for mitochondrial localization.

Type: Primary
Antigen: GST3/GST Pi
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat