Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-CD134/OX40 Polyclonal Antibody
  BSBTA02495
undefined
Anti-CD134/OX40 Polyclonal Antibody
  BSBTA02495

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody type:
    Primary
  • Antigen symbol:
    CD134/OX40
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Rat,
    Mouse
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Rabbit IgG polyclonal antibody for CD134/OX40 detection. Tested with WB, Direct ELISA in Mouse;Rat.

Type: Primary
Antigen: CD134/OX40
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse; Rat