Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Cathepsin B Polyclonal Antibody
Anti-Cathepsin B Polyclonal Antibody
  BSBTPB9046
undefined
Anti-Cathepsin B Polyclonal Antibody
  BSBTPB9046

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody type:
    Primary
  • Antigen name:
    Cathepsin B
  • Antigen symbol:
    CTSB
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Polyclonal antibody for Cathepsin B/CTSB detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. Cathepsin B/CTSB information: Molecular Weight: 37822 MW; Subcellular Localization: Lysosome. Melanosome. Secreted, extracellular space . Identified by mass spectrometry in melanosome fractions from stage I to stage IV.

Type: Primary
Antigen: Cathepsin B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human