
Medroxyprogesterone acetate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:71-58-9
:
Formula:C₂₄H₃₄O₄
MDL Number:MFCD00010483
:386.53 g/mol
Melting Pt:206…208 °C
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Melting point:203 to 209 °C
- Loss on drying:≤1 %
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together