Droperidol
Catalog # ACRO460840010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Loss on drying:≤5 % (70°C, 4 h) (vaccuum)
- Titration with HClO4:98 to 102 % (on dry substance)
Specification Test Results
About this item
:
Melting Pt:148…149 °C
:379.43 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:548-73-2
Formula:C₂₂H₂₂FN₃O₂
MDL Number:MFCD00083290