ContactUsLinkComponent
L-Lysine methyl ester dihydrochloride 98%
undefined
L-Lysine methyl ester dihydrochloride 98%
  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Specific optical rotation:
    +21° to +25° (20°C, 589 nm) (1% in MeOH)
  • Titration with HClO4:
    ≥97.5 %
Frequently Bought Together