ContactUsLinkComponent
N-Hexadecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-16, SB-16) 98%
Irritant
N-Hexadecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-16, SB-16) 98%
Certificates
undefined
Irritant
N-Hexadecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-16, SB-16) 98%
Specification Test Results
  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤2 %
  • HPLC:
    ≥97.5 %
  • Elemental Analysis:
    63.1 to 65.7 % (C, anhydrous)