
(S)-Tetrahydrofuran-3-amine hydrochloride 95%
Catalog # ACRO456930050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to brown Powder or solid
- Infrared spectrum:Conforms
- Titration Argentometric:≥94.0 to ≤106.0 %
- Specific optical rotation:-2° to -8° (20°C, 589 nm) c=0.5 in Methanol
Specification Test Results
About this item
:
:123.58 g/mol
Melting Pt:140…145 °C
:GHS07
Storage Temperature:Ambient
CAS Number:204512-95-8
Formula:C₄H₁₀ClNO
MDL Number:MFCD08445642