
4-[5-(4-Methylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulphonamide 98%
Catalog # ACRO456451000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Melting point:159 to 165 °C
- HPLC:≥97.5 %
Specification Test Results
About this item
:
:381.38 g/mol
Melting Pt:156…157 °C
:GHS08
Storage Temperature:Ambient
CAS Number:169590-42-5
Formula:C₁₇H₁₄F₃N₃O₂S
MDL Number:MFCD00941298