
Methyl hexanoate 99% analytical standard for GC
:106-70-7
:
:C₇H₁₄O₂
:MFCD00009510
:130.19 g/mol
:151 °C (760 mmHg)
:–71 °C
:0,884 g/cm³
:42 °C
:Ambient
:3272
:3,III
:GHS02
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4040 to 1.4060 (20°C, 589 nm)
- GC:≥98.5 %
Frequently Bought Together