
Vinylphosphonic acid 90%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1746-03-8
:
Formula:C₂H₅O₃P
MDL Number:MFCD00043866
:108.0331 g/mol
Melting Pt:36 °C
Density:1,37 g/cm³
Flash Pt:113 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Titration Acid:≥88 %
- Water:≤0.5 %
Specification Test Results
Frequently Bought Together