DNPH standards, organotin mixtures, CERTAN®
Catalog # PROOSL31005
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:10 mL
- Standards:Reference standard
- Description:Organotin mix 8 - stock solution, 1 mg/ml
Specifications
About this item
These selected organotin compounds are determined by gas chromatography using DIN 38407-F13.
Certifications: According to DIN EN ISO 17353.