Doxorubicin hydrochloride
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Doxorubicin HCl, Hydroxydaunorubicin Hydrochloride, (8S,10S)-10-(((2R,4S,5S,6S)-4-Amino-5-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-7,8,9,10-tetrahydrotetracene-5,12-dione hydrochloride
CAS Number:25316-40-9
:
Formula:C₂₇H₂₉NO₁₁·HCl
MDL Number:MFCD00077757
:579.98 g/mol
Merck Index:14,3439
Melting Pt:216 °C (dec.)
EINECS:246-818-3
:GHS08
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Certificates
Search by Lot NumberOrganic Building Blocks and Intermediates
Application Notes
Thermo Scientific Cell Culture Workflow
Product Literature
Frequently Bought Together