Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Wipes
Disposable Wipes and Towels
Paper Towels
Hand towels, C-fold, Pristine
Hand towels, C-fold, Pristine
Hand towels, C-fold, Pristine
Catalog # BUNZ067368
Supplier:
BUNZL
Expand image
Hand towels, C-fold, Pristine
Catalog # BUNZ067368
Supplier:
BUNZL
Specifications
Packed:
2400/case
Colour:
White
Description:
Pristine C-fold hand towel, 2 ply
L×W:
95×230 mm
Pk:
2.400
Specifications
About this item
Product Details
Product Details