7-Ethyl-10-hydroxycamptothecin 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:86639-52-3
:MFCD06762720
Formula:C₂₂H₂₀N₂O₅
MDL Number:MFCD00871873
:392.40 g/mol
Storage Temperature:Freezer
UN:1544
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Off-white to yellow Powder
- Infrared spectrum:Conforms
- Water:≤5 %
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together