Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-ORAI3 Mouse Monoclonal Antibody [clone: 1B4F1 ]
Anti-ORAI3 Mouse Monoclonal Antibody [clone: 1B4F1 ]
Catalog # PRSIPM-4913
Supplier:  ProSci Inc.
undefined
Anti-ORAI3 Mouse Monoclonal Antibody [clone: 1B4F1 ]
Catalog # PRSIPM-4913
Supplier:  ProSci Inc.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ORAI calcium release-activated calcium modulator 3
  • Antigen symbol:
    ORAI3
  • Clonality:
    Monoclonal
  • Clone:
    1B4F1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Format:
    Purified Antibody
  • Form:
    Liquid
  • Gene ID:
    93129
  • Antigen synonyms:
    ORAI3 Antibody [1B4F1] : TMEM142C
  • Storage temperature:
    ORAI3 monoclonal antibody can be stored at -20˚C, stable for one year.
  • Concentration:
    1 mg/mL
  • Shipping temperature:
    4°C
  • Immunogen:
    A 19 amino acid synthetic peptide from near the carboxy terminus of human ORAI3.
  • Purification:
    ORAI3 Monoclonal Antibody is immunoaffinity chromotography purified IgG.
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

ORAI3 Monoclonal Antibody: Antigen stimulation of immune cells triggers Ca++ entry t hrough Ca++ release-activated Ca++ (CRAC) channels. ORAI3 is one of two mammalian homologs to ORAI1, a recently identified four-transmembrane spanning protein that is an essential component of CRAC. All three homologs have been shown to function as Ca++ plasma membrane channels gated through interactions with STIM1, the store-activated endoplasmic reticulum Ca++ sensor. However, ORAI3 channels failed to produce detectable Ca++ selective currents in cells co-transfected with ORAI3 and STIM1, indicating that ORAI3 channels undergo a lesser degree of depotentiation than ORAI1 or ORAI2. Na+ currents through ORAI1, 2 and 3 channels were equally inhibited by extracellular Ca++, indicating that each have similar affinities for Ca++ within the selectivity filter. This antibody is predicted to have no cross-reactivity to ORAI1 or ORAI2. Larger molecular weight bands are sometimes seen in SDS-PAGE; these may represent post-translationally modified ORAI 3.

ORAI3 antibody can be used for detection of ORAI3 by Western blot at 2 μg/mL.

Type: Primary
Antigen: ORAI3
Clonality: Monoclonal
Clone: 1B4F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat