Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:endonuclease G
- Antigen symbol:ENDOG
- Clonality:Monoclonal
- Clone:7F2G10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human,Rat
- Western blot:Yes
- Format:Purified Antibody
- Form:Liquid
- Gene ID:2021
- Antigen synonyms:EndoG Antibody [7F2G10] : Endo G
- Storage temperature:EndoG monoclonal antibody can be stored at -20˚C, stable for one year.
- Concentration:1 mg/mL
- Shipping temperature:4°C
- Immunogen:Recombinant protein corresponding to amino acids 51 - 140 of human EndoG.
- Purification:EndoG Monoclonal Antibody is immunoaffinity chromotography purified IgG.
- Size:100 μg
- Pk:100 µG
Specifications
About this item
EndoG Monoclonal Antibody: The fragmentation of nuclear DNA is a hallmark of apoptotic cell death. The activities of caspase and nuclease are involved in the DNA fragmentation. Caspase-activated deoxyribonuclease (CAD), also termed DNA fragmentation factor (DFF40), is one such nuclease, and is capable of inducing DNA fragmentation and chromatin condensation after cleavage by caspase-3 of its inhibitor ICAD/DFF45. Caspase and CAD independent DNA fragmentation also exists. Recent studies demonstrated that another nuclease, endonuclease G (EndoG), is specifically activated by apoptotic stimuli and is able to induce nucleosomal fragmentation of DNA independently of caspase and DFF/CAD. EndoG is a mitochondrion-specific nuclease that translocates to the nucleus and cleaves chromatin DNA during apoptosis. The homologue of mammalian EndoG is the first mitochondrial protein identified to be involved in apoptosis in C. elegans. EndoG also cleaves DNA in vitro.
EndoG monoclonal monoclonal antibody can be used for detection of EndoG by Western blot at 5 - 10 μg/mL.
Type: Primary
Antigen: ENDOG
Clonality: Monoclonal
Clone: 7F2G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat