Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:BCL2 binding component 3
- Antigen symbol:BBC3
- Clonality:Monoclonal
- Clone:2A9G5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Format:Purified Antibody
- Form:Liquid
- Gene ID:27113
- Antigen synonyms:PUMA Antibody [2A9G5] : JFY1, PUMA, JFY-1
- Storage temperature:PUMA monoclonal antibody can be stored at -20˚C, stable for one year.
- Concentration:1 mg/mL
- Shipping temperature:4°C
- Immunogen:Recombinant protein corresponding to amino acids 76 - 170 of human PUMA-alpha.
- Purification:PUMA Monoclonal Antibody is immunoaffinity chromotography purified IgG.
- Size:100 μg
- Pk:100 µG
Specifications
About this item
PUMA Monoclonal Antibody: Apoptosis is related to many diseases and development. The p53 tumor-suppressor protein induces apoptosis through transcriptional activation of several genes. A novel p53 inducible pro-apoptotic gene was identified recently and designated PUMA (for p53 upregulated modulator of apoptosis) and bbc3 (for Bcl-2 binding component 3) in human and mouse. PUMA/bbc3 is one of the pro-apoptotic Bcl-2 family members including Bax and Noxa, which are also transcriptional targets of p53. The PUMA gene encodes two BH3 domain-containing proteins termed PUMAα and PUMAβ. PUMA proteins bind Bcl-2, localize to the mitochondria, and induce cytochrome c release and apoptosis in response to p53. PUMA may be a direct mediator of p53-induced apoptosis.
PUMA antibody can be used for detection of PUMA by Western blot at 2.5 to 5 μg/mL.
Type: Primary
Antigen: BBC3
Clonality: Monoclonal
Clone: 2A9G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat