Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
Leflunomide, Reference Standards

Leflunomide, Reference Standards

Supplier:   USP
Leflunomide, Reference Standards
Supplier:  USP
Certificates

4 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1356960.
  • 1357045.
  • 1357056.
  • 1357067.
  • Description:
  • Leflunomide
  • Leflunomide Related Compound A
  • Leflunomide Related Compound B
  • Leflunomide Related Compound C
  • Synonym(s):
  • 4-(trifluoromethyl)aniline
  • (Z)-2-cyano-3-hydroxy-N-(4-(trifluoromethyl)phenyl)but-2-enamide
  • N-(3'-Trifluoromethylphenyl)-5-methylisoxazole-4-carboxamide
  • CAS No.:
  • 75706-12-6
  • 455-14-1
  • 163451-81-8
  • 61643-23-0
  • Formula:
  • C₁₂H₉F₃N₂O₂
  • C₇H₆F₃N
  • C₁₂H₉F₃N₂O₂
  • C₁₂H₉F₃N₂O₂
  • UN No.:
  • 2811
  • 2810
  • 2811

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1356960.
    Leflunomide
    Leflunomide
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - £472.00
    £472.00
    1 * 200 mg
       £472.00

    Specifications:
    More
    Specifications
    Cat. No.
    1356960.
    Description
    Leflunomide
    Synonym(s)
    CAS No.
    75706-12-6
    Formula
    C₁₂H₉F₃N₂O₂
    UN No.
    2811
  • 1357045.
    Leflunomide Related Compound A
    Leflunomide Related Compound A
    USP reference standard
    Ampule
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - £1,160.00
    £1,160.00
    1 * 100 mg
       £1,160.00

    Specifications:
    More
    Specifications
    Cat. No.
    1357045.
    Description
    Leflunomide Related Compound A
    Synonym(s)
    4-(trifluoromethyl)aniline
    CAS No.
    455-14-1
    Formula
    C₇H₆F₃N
    UN No.
    2810
  • 1357056.
    Leflunomide Related Compound B
    Leflunomide Related Compound B
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - £1,240.00
    £1,240.00
    1 * 20 mg
       £1,240.00

    Specifications:
    More
    Specifications
    Cat. No.
    1357056.
    Description
    Leflunomide Related Compound B
    Synonym(s)
    (Z)-2-cyano-3-hydroxy-N-(4-(trifluoromethyl)phenyl)but-2-enamide
    CAS No.
    163451-81-8
    Formula
    C₁₂H₉F₃N₂O₂
    UN No.
    2811
  • 1357067.
    Leflunomide Related Compound C
    Leflunomide Related Compound C
    USP reference standard
    Vial
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - £1,070.00
    £1,070.00
    1 * 10 mg
       £1,070.00

    Specifications:
    More
    Specifications
    Cat. No.
    1357067.
    Description
    Leflunomide Related Compound C
    Synonym(s)
    N-(3'-Trifluoromethylphenyl)-5-methylisoxazole-4-carboxamide
    CAS No.
    61643-23-0
    Formula
    C₁₂H₉F₃N₂O₂
    UN No.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.