ContactUsLinkComponent
Elemental analysis standards, phosphate rock, western, NIST-694
Elemental analysis standards, phosphate rock, western, NIST-694
  PROONIST-694
 :  
Elemental analysis standards, phosphate rock, western, NIST-694
  PROONIST-694
 :  
  • Pk:
    90 g
  • Description:
    NIST-694, western phosphate rock
This standard material is intended to use in validating analytical methods and in calibrating instruments used in the analysis of phosphate rock, fertilisers and similar materials.
A unit consists of 90 g of <150 µm (100 mesh) powdered material.
 : Traceable to NIST and SRM.