Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Phosphomannomutase 1
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:30.8 kD
- Sequence:Met1-Ala262
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, 1 mM DTT, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Phosphomannomutase 1 (PMM1) blongs to the eukaryotic PMM family. Phosphomannomutase 1 can catalyses the conversion between D-mannose 6-phosphate and D-mannose 1-phosphate which is a substrate for GDP-mannose synthesis. GDP-mannose is used for synthesis of dolichol-phosphate-mannose which required for a number of critical mannosyl transfer reactions. PMM1 is highly expressed in liver, heart, brain, and pancreas, but lower expression in skeletal muscle. In addition, PMM1 may be responsible for the degradation of glucose-1,6 bisphosphate in ischemic brain.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.