Order Entry
Northern Ireland
ContactUsLinkComponent
Human recombinant IL22 (from CHO cells)
Human recombinant IL22 (from CHO cells)
Catalog # PRSI90-515
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant IL22 (from CHO cells)
Catalog # PRSI90-515
Supplier:  ProSci Inc.
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    25 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Tag sequence:
    Fc Tag
  • Storage conditions:
    Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Protein synonyms:
    IL-10-related T Cell-derived-inducible Factor|Interleukin-22
  • Protein/peptide name:
    IL22
  • Purity:
    >98% (SDS-PAGE)
  • Sequence:
    34-179
  • Formulation:
    Lyophilized from 0.2um-filtered solution in PBS.

Specifications

About this item

Interleukin-22 (IL-22), also known as IL-10 related T cell derived inducible factor (ILTIF) was initially identified as a gene induced by IL-9 in mouse T cells and mast cells. IL-22 has been shown to activate STAT1 and STAT3 in several hepatoma cell lines and upregulate the production of acute phase proteins. IL-22 is produced by normal T cells upon anti-CD3 stimulation in humans. Mouse IL-22 expression is also induced in various organs upon lipopolysaccharide injection, suggesting that IL-22 may be involved in inflammatory responses. The functional IL-22 receptor complex consists of two receptor subunits, IL-22R(CRF29) and IL-10Rbeta(CRF24), belonging to the class II cytokine receptor family.

Fusion-Tag: Fc Tag

This recombinant proteins is for research use only.