Order Entry
Northern Ireland
ContactUsLinkComponent
Human recombinant IL18 (from E. coli)
Human recombinant IL18 (from E. coli)
Catalog # PRSI90-506
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant IL18 (from E. coli)
Catalog # PRSI90-506
Supplier:  ProSci Inc.
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    His Tag
  • Storage conditions:
    Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Protein synonyms:
    Interferon-gamma-inducing Factor|IL1F4|Interleukin-18|IGIF|IL-1gamma
  • Protein/peptide name:
    IL18
  • Purity:
    >97% (SDS-PAGE)
  • Sequence:
    37-193
  • Formulation:
    Lyophilized from 0.2um-filtered solution in PBS.

Specifications

About this item

Interleukin-18 (IL-18) is a costimulatory factor for production of interferon-gamma (IFN-gamma) in response to toxic shock and shares functional similarities with IL-12. IL-18 is synthesised as a precursor 24kDa molecule without a signal peptide and must be cleaved to produce an active molecule. IL-1 converting enzyme (ICE; Caspase-1) cleaves pro-IL-18 at aspartic acid in the P1 position, producing the mature, bioactive peptide that is readily released from the cells. It is reported that IL-18 is produced from Kupffer cells, activated macrophages, keratinocytes, intestinal epithelial cells, osteoblasts, adrenal cortex cells and murine diencephalon. IFN-gamma is produced by activated T or NK cells and plays critical roles in the defense against microbiral pathogens. IFN-gamma activates macrophages and enhances NK activity and B cell maturation, proliferation and Ig secretion. IFN-gamma also induces expression of MHC class I and II antigens and inhibits osteoclast activation. IL-18 acts on T helper type-1 (Th1) T cells and in combination with IL-12 strongly induces them to produce IFN-gamma. Pleiotropic effects of IL-18 have also been reported, such as enhancement production of IFN-gamma and GM-CSF in peripheral blood mononuclear cells, production of Th1 cytokines, IL-2, GM-CSF, IFN-gamma in T cells and enhancement of Fas ligand expression by Th1 cells.

Fusion-Tag: His Tag

This recombinant proteins is for research use only.