Order Entry
Northern Ireland
ContactUsLinkComponent
Human recombinant IL12 (from CHO cells)
Human recombinant IL12 (from CHO cells)
Catalog # PRSI90-502
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant IL12 (from CHO cells)
Catalog # PRSI90-502
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    25 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Tag sequence:
    Fc Tag
  • Storage conditions:
    Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Protein synonyms:
    IL-12p70|NKSF|Cytotoxic Lymphocyte Maturation Factor|CLMF|TSF|T-cell Stimulatory Factor|IL-12p75|Interleukin-12|Natural Killer Cell Stimulatory Factor
  • Protein/peptide name:
    IL12
  • Purity:
    >98% (SDS-PAGE)
  • Sequence:
    57-253
  • Formulation:
    Lyophilized from 0.2um-filtered solution in PBS.

Specifications

About this item

Interleukin-12 (IL-12), also known as Natural Killer Cell Stimulatory Factor (NKSF) or Cytotoxic Lymphocyte Maturation Factor (CLMF), is a heterodimeric pleiotropic cytokine made up of a 40 kDa (p40) subunit and a 35 kDa (p35) subunit. IL-12 is produced by macrophages and B lymphocytes and has been shown to have multiple effects on T cells and Natural Killer (NK) cells. Some of these IL-12 activities include the induction of IFN-gamma and TNF in resting and activated T and NK cells; the enhancement of cytotoxic activity of resting NK and T cells, the stimulation of resting T cell proliferation in the presence of a comitogen; and the enhancement of NK cell proliferation. Current evidence indicates that IL-12 is a key mediator of cellular immunity and induces the differentiation of Th1 cells from precursor T helper cells. Based on its activities, it has been suggested that IL-12 may have therapeutic potential as a vaccine adjuvant that promotes cellular immunity and as an antitumour and anti viral agent.

Fusion-Tag: Fc Tag

This recombinant proteins is for research use only.