Order Entry
Northern Ireland
ContactUsLinkComponent
Human recombinant BAFF (from CHO cells)
Human recombinant BAFF (from CHO cells)
Catalog # PRSI90-301
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant BAFF (from CHO cells)
Catalog # PRSI90-301
Supplier:  ProSci Inc.
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Tag sequence:
    Fc Tag
  • Storage conditions:
    Stable for at least 6 months after receipt when stored at −20 °C.
  • Biological activity:
    Rescues the production of mature follicular and marginal zone B cells. - in vitro: Addition of 1-100 ng/ml of Fc-BAFF induces B cell survival in a dose-dependent manner. - in vivo: Fc-BAFF treatment rescues mature B cell development in BAFF deficient mice. Mice are injected at day 0 with 100 ug Fc-BAFF i.v. and at day 14 with 50 ug Fc-BAFF i.p.
  • Protein synonyms:
    B Cell Activating Factor|CD257|TALL-1|BLyS|Tumor Necrosis Factor Ligand Superfamily Member 13B|TNFSF13B
  • Protein/peptide name:
    BAFF
  • Purity:
    >95% (SDS-PAGE)
  • Sequence:
    136-285
  • Formulation:
    Lyophilized, Contains PBS.

Specifications

About this item

BAFF is mainly produced by innate immune cells such as neutrophils, monocytes, macrophages, dendritic cells, follicular dendritic cells. T cells, activated B cells, some malignant B cells and also non-lymphoid cells like astrocytes, synoviocytes and epithelial cells can also produce BAFF. BAFF binds three distinct receptors (BAFF-R, TACI and BCMA) expressed predominantly on B cells, although activated T cells also express BAFF-R. BAFF is a master regulator of peripheral B cell survival, and together with IL-6, promotes Ig class-switching and plasma cell differentiation. Besides its major role in B cell biology, BAFF co-stimulates activated T cells. Deregulated expression of BAFF leads to autoimmune disorders in mice. In humans, elevated levels of soluble BAFF have been detected in the serum of patients with various autoimmune diseases such as Sjoegren syndrome, Rheumatoid arthritis (RA), Multiple sclerosis (MS) and Systemic Lupus Erythematosus (SLE). BAFF has also increased levels in some lymphoid cancers.

Fusion-Tag: Fc Tag

This recombinant proteins is for research use only.