Order Entry
Northern Ireland
ContactUsLinkComponent
Human recombinant EDAR (from CHO cells)
Human recombinant EDAR (from CHO cells)
Catalog # PRSI90-297
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant EDAR (from CHO cells)
Catalog # PRSI90-297
Supplier:  ProSci Inc.
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Tag sequence:
    Fc Tag
  • Storage conditions:
    Stable for at least 6 months after receipt when stored at −20 °C.
  • Biological activity:
    Inhibits EDA-A1 activity.
  • Protein synonyms:
    Ectodermal Dysplasia Receptor|Ectodysplasin-A Receptor|Anhidrotic Ectodysplasin Receptor 1|Tumor Necrosis Factor Receptor Superfamily Member EDAR|Downless Homolog|EDA-A1 Receptor
  • Protein/peptide name:
    EDAR
  • Purity:
    >95% (SDS-PAGE)
  • Sequence:
    27-183
  • Formulation:
    Lyophilized, Contains PBS.

Specifications

About this item

The TNF family ligand ectodysplasin A (EDA) and its receptor EDAR are required for proper development of skin appendages such as hair, teeth, and eccrine sweat glands. Loss of function mutations in the Eda gene cause X-linked hypohidrotic ectodermal dysplasia (XLHED), a condition that can be ameliorated in mice and dogs by timely administration of recombinant EDA. The Eda gene on the X chromosome is transcribed as multiple splice variants, only two of which code for the receptor-binding C-terminal TNF homology domain. These two variants code for 391- and 389-amino acid-long proteins called EDA1 and EDA2. EDA1 binds EDAR, whereas EDA2 binds to another receptor, XEDAR. The biology of EDA2 and XEDAR is distinct from that of EDA1. Indeed, XEDAR-deficient mice have no obvious ectodermal dysplasia phenotype, whereas mice deficient in EDA, EDAR, or the signalling adaptor protein EDARADD all display virtually indistinguishable ectodermal dysplasia phenotypes, indicating the predominance of the EDA1-EDAR axis in the development of skin-derived appendages.

Fusion-Tag: Fc Tag

This recombinant proteins is for research use only.