Order Entry
Northern Ireland
ContactUsLinkComponent
 
Human recombinant Annexin V (from E. coli)
Catalog # PRSI90-218
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Annexin V (from E. coli)
Catalog # PRSI90-218
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    GST tag
  • Storage conditions:
    Stable for at least 1 year after receipt when stored at −20 °C. Reconstituted solutions can be kept at 4 °C under sterile conditions for one week or should be frozen in aliquots at −20 °C.
  • Protein synonyms:
    Endonexin II|Calphobindin I|Placental Anticoagulant Protein I|Vascular Anticoagulant-alpha|Lipocortin V|Annexin A5
  • Protein/peptide name:
    Annexin V
  • Purity:
    >95% (SDS-PAGE)
  • Formulation:
    Lyophilized, In PBS, pH 7.4.

Specifications

About this item

Annexin V is an anticoagulant protein that acts as an indirect inhibitor of the thromboplastin-specific complex, which is involved in the blood coagulation cascade. Annexin V competes for phosphatidylserine binding sites (e. g. platelets) with prothrombin or coagulation factors and inhibits the activity of phospholipase A1. Antibodies directed against annexin V are found in patients with a disease called the antiphospholipid syndrome(APS), a thrombophilic disease associated with autoantibodies against phospholipid compounds. Annexin V is used as a probe in the 'annexin V affinity assay' to detect cells that have expressed phosphatidylserine on the cell surface, a feature found in apoptosis and other forms of cell death.

Fusion-Tag: GST tag

This recombinant proteins is for research use only.