Order Entry
Northern Ireland
ContactUsLinkComponent
Mouse recombinant complement factor D
Mouse recombinant complement factor D
Catalog # PRSI92-692
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant complement factor D
Catalog # PRSI92-692
Supplier:  ProSci Inc.
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Tag sequence:
    C-10 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Df|Properdin factor D|28 kDa adipocyte protein|Complement factor D|C3 convertase activator|Adn|Adipsin|Cfd
  • Protein/peptide name:
    Complement factor D
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    26.8 kD
  • Sequence:
    Ile26-Ser259
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mMTris,150mMNaCl,pH7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

Specifications

About this item

Complement factor D, also known as adipsin, is a member of the chymotrypsin family of serine proteases, which plays an essential role in host defence as the rate-limiting enzyme in the alternative pathway of complement activation. Complement factor D activates a convertase (C3bBb) responsible for cleavage of the complement protein C3, which leads to the activation of terminal complement component C5-9 to form the membrane attack complex on microbial or cellular surfaces. It also functions in the regulation of systemic energy balance and physiologic and pathologic processes, including immunity and inflammation.

Fusion-Tag: C-10 His tag

This recombinant protein can be used for biological assays. For research use only.