Order Entry
Northern Ireland
ContactUsLinkComponent
Mouse recombinant decorin
Mouse recombinant decorin
  PRSI92-678
 :  
Mouse recombinant decorin
  PRSI92-678
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Decorin| PG-S2|PG40| PG40| Bone proteoglycan II|PGS2| Dcn
  • Protein/peptide name:
    Decorin
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    39 kD
  • Sequence:
    Gly17-Lys354
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

 

 

Decorin, also known as PG40 and DCN, is a member of the class I family of small leucine-rich proteoglycans (SLRPs) that is expressed in the stroma of various forms of cancer and has been recently proposed to act as a guardian from the matrix. Mature human Decorin contains 12 tandem LRR and shares 80% and 78% aa sequence identity with mouse and rat Decorin, respectively. Decorin embraces numerous functions including: regulation of collagen fibrillogenic, hepatic carcinogenesis, fetal membrane and calcium homeostasis, keratinocyte function, and suppression of angiogenesis. Most recently, soluble decorin has been shown to induce autophagy in endothelial cells and mitophagy in breast carcinoma cells.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.