Mouse recombinant ectodysplasin receptor
Product Details & Documents
Ectodysplasin A receptor (EDAR) is a type I transmembrane protein of the TNF- alpha receptor superfamily which plays a key role in ectodermal differentiation. EDAR was encoded by the mouse down less gene and defective in human dominant and recessive forms of autosomal hypo hidrotic ectodermal dysplasia (EDA) syndrome. The extracellular domain of EDAR contains 14 cysteine residues, six of which approximate the TNFRSF cysteine-rich region, the cytoplasmic domain contains a region with homology to the death domains found in other TNFRSF members. EDAR has been suggested to be an early and important promoter of placode development in all ectodermal organs, such as uch as hair follicles, teeth and sweat glands. EDA-A1, the A1 isoform of EDA, is the EDAR ligand. EDA and EDA are implicated in appendage development by the cloning of a gene underlying hypo hidrotic ectodermal dysplasia (HED) in mouse and human. HED is Characterised by agenesis or malformation of ectoderm-derived appendages, such as teeth, sweat glands and hair follicles, while the skin itself develops normally.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Documents
75791-702.pdf
Datasheet
Specifications
Specifications
- Pk:50 µG
- Cat. No.:PRSI92-570
- Protein/peptide type:Recombinant
- Protein/peptide name:Ectodysplasin Receptor
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Formulation:Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Protein synonyms:EDAR|Anhidrotic ectodysplasin receptor 1|Tumor necrosis factor receptor superfamily member EDAR|Downless|Ectodysplasin-A receptor|Ectodermal dysplasia receptor
- Purity:> 95% as determined by reducing SDS-PAGE
- Tag sequence:C-Fc tag
- Sequence:Glu27-Ile189
- Species:Mouse
- Molecular weight:44.7 kD
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



