Order Entry
Northern Ireland
ContactUsLinkComponent
Human recombinant Methyl-CpG-binding 2 (from cells)
Human recombinant Methyl-CpG-binding 2 (from cells)
Catalog # PRSI92-318
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Methyl-CpG-binding 2 (from cells)
Catalog # PRSI92-318
Supplier:  ProSci Inc.
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    MECP2|Methyl-CpG-binding protein 2|MeCp-2 protein
  • Protein/peptide name:
    Methyl-CpG-binding 2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    53.5 kD
  • Sequence:
    Met1-Ser486
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM Tris,150mM NaCl,pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

The MeCP2 helps regulate gene activity (expression) by modifying chromatin, the complex of DNA and protein that packages DNA into chromosomes. The MeCP2 protein is present in cells throughout the body, although it is particularly abundant in brain cells.In the brain, the MeCP2 protein likely plays a role in maintaining connections (synapses) between neurons, where cell-to-cell communication occurs. The alternative splicing of proteins is critical for normal communication between neurons and may also be necessary for the function of other types of brain cells.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.