Order Entry
Northern Ireland
ContactUsLinkComponent
Human recombinant FAM19A4 (from E. coli)
Human recombinant FAM19A4 (from E. coli)
Catalog # PRSI92-258
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant FAM19A4 (from E. coli)
Catalog # PRSI92-258
Supplier:  ProSci Inc.
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    FAM19A4|Protein FAM19A4|chemokine-like protein TAFA-4|member A4|TAFA4|family with sequence similarity 19 (chemokine (C-C motif)-like)|Chemokine-like protein TAFA-4
  • Protein/peptide name:
    FAM19A4
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    14.1 kD
  • Sequence:
    Ser35-Arg140
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

FAM19A4 is a secreted, 12 kDa member of the FAM19/TAFA family of chemokine-like proteins. Like other members of the FAM19/TAFA family, with the exception of TAFA5, mature FAM19A4 contains 10 regularly spaced cysteine residues. The FAM19A4 proteins are predominantly expressed in specific regions of the brain and the biological functions of FAM19A4 family members remain to be determined, but there are a few tentative hypotheses. First, FAM19A4 may modulate immune responses in the CNS by functioning as brain specific chemokines, and may act with other chemokines to optimize the recruitment and activity of immune cells in the CNS. Second, FAM19A4 may represent a novel class of neurokines that act as regulators of immune nervous cells. And third, FAM19A4 may control axonal sprouting following brain injury.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.