Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:N-GST tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:CALC2|CALCB|Beta-type CGRP|CALCB.|Calcitonin gene-related Peptide II|Calcitonin gene-related Peptide 2
- Protein/peptide name:Calcitonin gene-related Peptide 2
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:40.9 kD
- Sequence:Met1-Ala127
- Formulation:Supplied as a 0.2 um filtered solution of PBS, pH 7.4, 50% glycerol. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
CALCB is a member of the calcitonin family. CALCB is produced in both peripheral and central neurons. It is a potent peptide vasodilator and can function in the transmission of pain. In the spinal cord, the function and expression of CGRP may differ depending on the location of synthesis. CALCB is derived mainly from the cell bodies of motor neurons when synthesised in the ventral horn of the spinal cord and may contribute to the regeneration of nervous tissue after injury.
Fusion-Tag: N-GST tag
This recombinant protein can be used for biological assays. For research use only.