Order Entry
Northern Ireland
ContactUsLinkComponent
Human recombinant FKBP52 (from E. coli)
Human recombinant FKBP52 (from E. coli)
Catalog # PRSI92-198
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant FKBP52 (from E. coli)
Catalog # PRSI92-198
Supplier:  ProSci Inc.
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    59kDa immunophilin|HSP-binding immunophilin|FK506-binding protein 4|Peptidyl-prolyl cis-trans isomerase FKBP4|FKBP59|Rotamase|51kDa FK506-binding protein|immunophilin FKBP52|FKBP4
  • Protein/peptide name:
    FKBP52
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    52.8 kD
  • Sequence:
    Met1-Ala459
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM Tris,10%glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

FKBP4 act as a regulator of microtubule dynamics by inhibiting MAPT/TAU ability to promote microtubule assembly. FKBP4 may play a role in the intracellular trafficking of heterooligomeric forms of steroid hormone receptors between cytoplasm and nuclear compartments, it also may have a protective role against oxidative stress in mitochondria. The isomerase activity controls neuronal growth cones via regulation of TRPC1 channel opening.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.