Order Entry
Northern Ireland
ContactUsLinkComponent
Human recombinant SF-20 (from E. coli)
Human recombinant SF-20 (from E. coli)
Catalog # PRSI92-191
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant SF-20 (from E. coli)
Catalog # PRSI92-191
Supplier:  ProSci Inc.
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    UPF0556 protein C19orf10|IL-25|C19orf10.|MYDGF|stromal cell-derived growth factor SF20|interleukin-25
  • Protein/peptide name:
    SF-20
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    18 kD
  • Sequence:
    Ser33-Leu173
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

C19orf10 is a secreted protein which belongs to the UPF0556 family. It is expressed in synovial tissue and detected in synovial fluid of patients with arthropaties. C19orf10 plays a role in proliferation of lymphoid cells and is considered an interleukin.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.