Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:IL-25|Interleukin-25|IL17E|IL25|Interleukin-17E|IL-17E
- Protein/peptide name:IL25
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:16.7 kD
- Sequence:Tyr33-Gly177
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM Tris,150mM NaCl 1mM EDTA,pH8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Interleukin 25 (IL-25) belongs to the Interleukin 17 (IL-17) family of proteins, which is comprised of six members (IL-17, IL-17B through IL-17F). These proteins are secreted and are structurally related by sharing a conserved cysteine-knot fold near the C-terminus, but have considerable sequence divergence at the N-terminus. With the exception of IL-17B, which exists as a non-covalently linked dimer, all IL-17 family members are disulfide-linked dimers. IL-17 family proteins are pro-inflammatory cytokines that induce local cytokine production and are involved in the regulation of immune functions. Human interleukin-17E (IL17E), also referred to as Interleukin-25 (IL25), is a distinct member of the IL17 cytokine family comprised of at least six members sharing a conserved cysteine-knot structure but divergent at the N-terminus. IL25 is a glycoprotein secreted as dimers by innate effector eosinophils and basophils, and present at very low levels in various peripheral tissues. IL25, together with IL17B, are ligands for the cytokine receptor IL17BR, and the cross-linking induces NF-κB activation and production of the proinflammatory chemokine IL-8, as well as ERK, JNK, and p38 activation. Overexpression of IL25 gene in transgenic mice suggested that this cytokine can regulate hematopoietic and immune functions, and additionally is identified as a proinflammatory cytokine favoring Th2-type immune responses possibly by enhancing the maintenance and functions of adaptive Th2 memory cells.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag