Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:IL-20RA|IL20RA|CRF2-8|IL-20R-Alpha|IL-20R1|IL-20 Receptor Subunit Alpha|Interleukin-20 Receptor Subunit Alpha|Cytokine Receptor Family 2 Member 8|Cytokine Receptor Class-II Member 8|ZcytoR7
- Protein/peptide name:IL-20 R alpha
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:26.3 kD
- Sequence:Val30-Lys250
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Interleukin-20 Receptor Subunit alpha (IL20RA) is a single-pass type I membrane protein that is a member of the type II cytokine receptor family. IL20RA is synthetized a 553 amino acid glycoprotein precursor containing a 29 amino acid signal peptide, a 221 amino acid extracellular domain with two fibronectin type-III domains, a 24 amino acid transmembrane region, and a 279 amino acid intracellular domain. IL20RA is widely expressed with highest levels found in skin and testis and high levels in brain. IL20RA forms a heterodimer with IL20RB, and the complex serves as a receptor for IL19, IL20 and IL24. IL20RA also forms a heterodimer with the unique and specific receptor IL10RB and functions as the receptor for IL26.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag