Order Entry
Northern Ireland
ContactUsLinkComponent
Human recombinant Fc-gamma RIIb (from cells)
Human recombinant Fc-gamma RIIb (from cells)
Catalog # PRSI91-400
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Fc-gamma RIIb (from cells)
Catalog # PRSI91-400
Supplier:  ProSci Inc.
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    CD32|IgG Fc Receptor II-b|Fc-Gamma-RIIb|Low Affinity Immunoglobulin Gamma Fc Region Receptor II-b|IGFR2|CD32b|FCG2|Fc-Gamma RII-b|CDw32|FCGR2B|FcRII-b
  • Protein/peptide name:
    Fc-gamma RIIb
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    20.64 kD
  • Sequence:
    Thr43-Pro217
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Fc gamma RIIB is a low affinity receptor that recognizes the Fc portion of IgG. The human CD32 group consists of Fc gamma RIIA, Fc gamma RIIB, and Fc gamma RIIC proteins that share 94-99% sequence identity in their extracellular domains but differ substantially in their transmembrane and cytoplasmic domains. Fc gamma RII protein is expressed on cells of both myeloid and lymphoid lineages as well as on cells of non-hematopoietic origin. Fc gamma RIIB has an intrinsic cytoplasmic immunoreceptor tyrosine-based inhibitory motif (ITIM) and delivers an inhibitory signal upon ligand binding. Ligation of Fc gamma RIIB on B cells down-regulates antibody production and in some circumstances may promote apoptosis. Co-ligation of Fc gamma RIIB on dendritic cells inhibits maturation and blocks cell activation. Fc gamma RIIB may also be a target for monoclonal antibody therapy for malignancies. Fc gamma RIIB plays an important negative-regulating role through modulating the signals from activation receptors.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag